Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TEM7/PLXDC1 Antibody (104), DyLight 488 , Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289972G
Description
TEM7/PLXDC1 Monoclonal antibody specifically detects TEM7/PLXDC1 in Human samples. It is validated for ELISASpecifications
TEM7/PLXDC1 | |
Monoclonal | |
DyLight 488 | |
FLJ45632, plexin domain containing 1,2410003I07Rik, plexin domain-containing protein 1, TEM3FLJ36270, TEM7DKFZp686F0937, Tumor endothelial marker 3, Tumor endothelial marker 7 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human TEM7/PLXDC1 (AAH36059.1; Met 1-Thr 426). | |
0.1 mL | |
Angiogenesis | |
57125 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
104 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction