Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TEM7/PLXDC1 Antibody (197C193 (IM193)) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 8 publications
$206.50 - $499.50
Specifications
Antigen | TEM7/PLXDC1 |
---|---|
Clone | 197C193 (IM193) |
Concentration | 1.0 mg/ml |
Dilution | Western Blot, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500. Use reported in scientific literature (Meng et al (2007)), Immunoprecipitation 1:10-1:500. Use reported in scientific literature (Nanda et al (2004)), Immunohistochemistry-Paraffin 2-5 ug/ml, Immunohistochemistry-Frozen reported in scientific literature (Lee et al (2006)) |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10056557T
![]() |
Novus Biologicals
NB10056557SS |
0.025 mg |
Each for $206.50
|
|
|||||
NB10056557
![]() |
Novus Biologicals
NB10056557 |
0.1 mg |
Each for $499.50
|
|
|||||
Description
TEM7/PLXDC1 Monoclonal specifically detects TEM7/PLXDC1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
TEM7/PLXDC1 | |
1.0 mg/ml | |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Angiogenesis | |
Q8IUK5 | |
57125 | |
Amino acids 409-425 (LQNNLSPKTKGTPVHLG) of human TEM7 were used to develop this monoclonal antibody. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
197C193 (IM193) | |
Western Blot, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500. Use reported in scientific literature (Meng et al (2007)), Immunoprecipitation 1:10-1:500. Use reported in scientific literature (Nanda et al (2004)), Immunohistochemistry-Paraffin 2-5 ug/ml, Immunohistochemistry-Frozen reported in scientific literature (Lee et al (2006)) | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
FLJ45632, plexin domain containing 1,2410003I07Rik, plexin domain-containing protein 1, TEM3FLJ36270, TEM7DKFZp686F0937, Tumor endothelial marker 3, Tumor endothelial marker 7 | |
PLXDC1 | |
IgG1 | |
Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title