Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TEM7/PLXDC1 Antibody, DyLight 550, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299869R
Description
TEM7/PLXDC1 Polyclonal specifically detects TEM7/PLXDC1 in Human samples. It is validated for ELISA.Specifications
TEM7/PLXDC1 | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
DyLight 550 | |
FLJ45632, plexin domain containing 1,2410003I07Rik, plexin domain-containing protein 1, TEM3FLJ36270, TEM7DKFZp686F0937, Tumor endothelial marker 3, Tumor endothelial marker 7 | |
Produced in rabbits immunized with purified, recombinant TEM7/PLXDC1 (AAH36059.1; Met1-Thr426). | |
0.1 mL | |
Angiogenesis | |
57125 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction