Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TEM7 Recombinant Rabbit Monoclonal Antibody (104), Invitrogen™

Catalog No. PIMA543428
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA543428 50 μL
PIMA543134 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA543428 Supplier Invitrogen™ Supplier No. MA543428
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Specificity: Human TEM7/PLXDC1

Recently, using SAGE (Serial Analysis of Gene Expression) technology, St. Croix et al, have identified 46 genes, whose expression is specifically elevated in tumor-associated endothelium. Nine of these genes were prominently expressed only in tumor endothelial cells (EC), but were absent or barely detectable in normal ECs, and named as Tumor Endothelial Markers (TEMs, TEM 1-9). TEM7 (Tumor endothelial marker 7) transcripts are specifically expressed in the endothelium of colorectal cancer, primary cancers of lung, pancreas, breast, and brain. TEM7 is expressed specifically in endothelium of these cancers, whether primary or metastasis. The other six members of this family (TEM1, 3, 4, 5, 8, and 9) also show similar expression pattern in lung and brain tumors, and liver metastasis. Since most of the genes expressed differentially in tumor endothelium are also expressed during angiogenesis, these newly discovered genes might provide important resources for basic and clinical studies of human angiogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TEM7
Applications ELISA
Classification Recombinant Monoclonal
Clone 104
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene PLXDC1
Gene Accession No. Q8IUK5
Gene Alias 2410003I07Rik; ADP-ribosylation factor-like 12; AI848450; Arl12; DKFZp686F0937; FLJ36270; FLJ45632; plexin domain containing 1; plexin domain-containing protein 1; PLXDC1; TEM3; TEM7; TEM7 related; tumor endothelial marker 3; Tumor endothelial marker 7; tumor endothelial marker7
Gene Symbols PLXDC1
Host Species Rabbit
Immunogen Recombinant Human TEM7/PLXDC1 protein.
Purification Method Protein A
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57125
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.