Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TEM8/ANTXR1 Antibody (200C1339(SB20)) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$548.00
Specifications
Antigen | TEM8/ANTXR1 |
---|---|
Clone | 200C1339(SB20) |
Concentration | 0.5 mg/mL |
Dilution | Western Blot 1-3 ug/ml, Immunohistochemistry 2-5 ug/ml, Immunocytochemistry/Immunofluorescence reported in scientific literature (Matsuda (2010)), Immunohistochemistry-Paraffin 2-5 ug/ml. Use reported in scientific literature (Hotchkiss et al (2005)) |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Description
TEM8/ANTXR1 Monoclonal specifically detects TEM8/ANTXR1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
TEM8/ANTXR1 | |
0.5 mg/mL | |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Angiogenesis, Cancer, Cellular Markers, Extracellular Matrix, Phospho Specific | |
Q9H6X2 | |
84168 | |
This monoclonal antibody was developed by immunizing mice with a synthetic peptide containing amino acids 481-497 (CINFTRVKNNQPAKYPL) of human TEM8. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
200C1339(SB20) | |
Western Blot 1-3 ug/ml, Immunohistochemistry 2-5 ug/ml, Immunocytochemistry/Immunofluorescence reported in scientific literature (Matsuda (2010)), Immunohistochemistry-Paraffin 2-5 ug/ml. Use reported in scientific literature (Hotchkiss et al (2005)) | |
Monoclonal | |
Purified | |
RUO | |
Human | |
anthrax toxin receptor 1, ATR2310008J16Rik, FLJ10601, FLJ11298, FLJ21776, TEM82810405N18Rik, Tumor endothelial marker 8 | |
ANTXR1 | |
IgG1 | |
Protein G purified | |
Nanda et al (2004) note a passage dependent loss of TEM8 in HUVEC suggesting that that TEM8 may be a specific marker of the endothelial cell (EC) phenotype which is lost as primary EC's age in culture and de-differentiate. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title