Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TEM8/ANTXR1 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$201.00 - $423.00
Specifications
Antigen | TEM8/ANTXR1 |
---|---|
Concentration | 1 mg/ml |
Dilution | Western Blot 0.5-2 ug/ml, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 2-10 ug/ml |
Applications | Western Blot, ELISA, Immunofluorescence |
Classification | Polyclonal |
Description
TEM8/ANTXR1 Polyclonal antibody specifically detects TEM8/ANTXR1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, ImmunofluorescenceSpecifications
TEM8/ANTXR1 | |
Western Blot 0.5-2 ug/ml, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 2-10 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
NP_444262 | |
84168 | |
IgG | |
Ion exchange chromatography |
1 mg/ml | |
Western Blot, ELISA, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Angiogenesis, Cellular Markers, Extracellular Matrix, Phospho Specific | |
PBS | |
anthrax toxin receptor 1, ATR2310008J16Rik, FLJ10601, FLJ11298, FLJ21776, TEM82810405N18Rik, Tumor endothelial marker 8 | |
Antibody was raised against a peptide corresponding to 13 amino acids near the C-terminus of human TEM8/ANTXR1. The immunogen is located within the last 50 amino acids.. Amino Acid Squence: RAPPPSRPPPRPSV | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title