Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TEM8/ANTXR1 Antibody, DyLight 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP321501V
Description
TEM8/ANTXR1 Polyclonal antibody specifically detects TEM8/ANTXR1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, ImmunofluorescenceSpecifications
TEM8/ANTXR1 | |
Polyclonal | |
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
anthrax toxin receptor 1, ATR2310008J16Rik, FLJ10601, FLJ11298, FLJ21776, TEM82810405N18Rik, Tumor endothelial marker 8 | |
Antibody was raised against a peptide corresponding to 13 amino acids near the C-terminus of human TEM8/ANTXR1. The immunogen is located within the last 50 amino acids.. Amino Acid Squence: RAPPPSRPPPRPSV | |
0.1 mL | |
Angiogenesis, Cancer, Cellular Markers, Extracellular Matrix, Phospho Specific | |
84168 | |
Store at 4C in the dark. |
Western Blot, ELISA, Immunofluorescence | |
DyLight 405 | |
50mM Sodium Borate | |
Rabbit | |
Ion exchange chromatography | |
RUO | |
Primary | |
Human, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction