Learn More
BINDER™ Temperature and Humidity Calibration Certificate
Calibration certificate including 27 temperature measurements and 1 humidity measurement.
Supplier: BINDER™ 80122578
Description
- 27 temperature measuring points
- 1 humidity measuring point, at 25°C/60% RH, or at specified values
Specifications
| KBF PRO Series | |
| DIN12880 | |
| 1 Day | |
| Calibration Certificate |