Learn More
BINDER™ Temperature and Humidity Calibration Certificate
SDP
Calibration certificate including 27 temperature measurements and 1 humidity measurement.
$3602.00
Specifications
Certifications/Compliance DIN12880
Duration 1 Day
Product Type Calibration Certificate
View More Specs
Products 2
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity & Availability  
50-287-5768
SDP
View Documents
BINDER™
80122577
KBF Series
Each for $3,602.00
Only null left
Add to Cart
50-287-5726
SDP
View Documents
BINDER™
80122578
KBF PRO Series
Each for $3,602.00
Only null left
Add to Cart
Description
Description
  • 27 temperature measuring points
  • 1 humidity measuring point, at 25°C/60% RH, or at specified values
Specifications
Specifications
DIN12880
1 Day
Calibration Certificate
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.