Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BINDER™ Temperature Sensor
Shop All BINDER Inc. Products

Click to view available options
For Use With (Equipment):
Units Series M
Description
- Pt100 temperature sensor
- Additional flexible Pt100 for displaying the temperature on the unit display
Specifications
Specifications
Description | Pt100 Temperature Sensor |
For Use With (Equipment) | Units Series M |
Material | Plastic |
Weight (English) | 2.20 lb. |
Weight (Metric) | 1 kg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction