Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Tempol
GREENER_CHOICESDP

Catalog No. 3082100 Shop All Novus Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
30-82100 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 30-82100 Supplier R&D Systems™ Supplier No. 3082/100
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Superoxide scavenger; antioxidant

Tempol is a superoxide scavenger that displays neuroprotective, anti-inflammatory and analgesic effects. Tempol reduces γ-radiation-induced damage in hematopoietic stem cells in vitro and in vivo.

Chemical Identifiers

CAS 2226-96-2
Molecular Formula C9H18NO2

Specifications

Quantity 100 mg
Product Line Tocris
Formula Weight 172.24
Purity >99%
Chemical Name or Material Tempol
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.