Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tempshield™ Cryo-Gloves™ Pink Mid-Arm Length Cryogenic Gloves
SDP

Catalog No. 19230263 Shop All Tempshield Inc. Products
Click to view available options
Size:
Small
Medium
Large
X-Large
This item is not returnable. View return policy
Not for immersion in liquid nitrogen, for use near open flame, or in hazardous areas where ignition could occur.
This item is not returnable. View return policy
Not for immersion in liquid nitrogen, for use near open flame, or in hazardous areas where ignition could occur.

State-of-the art materials and multi-layered construction provide cryogenic protection for ultra-cold applications down to -196°C (-320°F).

State-of-the art materials and multi-layered construction provides cryogenic protection for ultra-cold applications down to -196°C (-320°F).

  • Cryogenic protection for ultra-cold applications down to -196°C (-320°F)
  • State-of-the art materials and multi-layered construction allow for a maximum level of thermal protection, flexibility, and dexterity
  • High performance, thermal lining wicks moisture away from hands to maintain comfort over extended periods of wear
  • Ideal for applications where function is important and safety is critical
  • Not intended for immersion in liquid nitrogen or other cryogenic liquids

Specifications

Certifications/Compliance (Europe) EN 420, EN 511
Description Mid-Arm Length
Color Pink
Hand Specific
Product Type Cryogenic Gloves
Size Large
EN388 Standard Yes
For Use With (Application) Dry Ice Handling, Low/Ultra-Low Temperature Freezers, and Closed Cryogenic Systems
Series Cryo-Gloves™
Temperature (English) Min -320°F
Temperature (Metric) Min. -196°C
Length (English) 13.3 to 15.5 in.
Length (Metric) 33.5 to 39.5 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.