Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Temsirolimus
GREENER_CHOICESDP

Catalog No. 526410 Shop All R&D Systems Products
Change view
Click to view available options
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
52-641-0 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 52-641-0 Supplier Tocris Bioscience™ Supplier No. 5264/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

mTOR inhibitor; antitumor

Temsirolimus is a mTOR inhibitor; inhibits tumor growth in breast cancer cell lines (IC50 values are 1.6 and 4.3 nM for SKBr3 and BT474, respectively). Inhibits HIF-1α-mediated VEGF production in breast cancer cell lines (BT474 and MDA-MB-231). Directly inhibits serum and VEGF mediated endothelial cell proliferation and morphogenesis in vitro and vessel formation in vivo. Causes G1/S cell cycle arrest in multiple cancer cell lines. Antiangiogenic. Activates autophagy.

Specifications

Quantity 10 mg
Product Type Temsirolimus
Purity >98%
Molecular Weight (g/mol) 1030.29
Inhibitors Temsirolimus

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.