Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tenascin C Antibody (4C8MS), Allophycocyanin/Cy7, Novus Biologicals™
SDP

Catalog No. N1068136AP7 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N1068136AP7 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N1068136AP7 Supplier Novus Biologicals Supplier No. NB11068136APCCY7
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Tenascin C Monoclonal specifically detects Tenascin C in Human, Mouse, Rat, Feline samples. It is validated for Flow Cytometry.

Specifications

Antigen Tenascin C
Applications Flow Cytometry
Classification Monoclonal
Clone 4C8MS
Conjugate APC-Cyanine7
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias 150-225, Cytotactin, Glioma-associated-extracellular matrix antigen, GMEM, GP 150-225, Hexabrachion, hexabrachion (tenascin C, cytotactin), hexabrachion (tenascin), HXBcytotactin, JI, MGC167029, Myotendinous antigen, neuronectin, tenascin, tenascin C, Tenascin-C, tenascin-C isoform 14/AD1/16, TN-C, TNGP
Gene Symbols TNC
Host Species Mouse
Immunogen Recombinant human Tenascin C [Swiss-Prot No. P24821].
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Breast Cancer, Cancer, Extracellular Matrix, Neuroscience, Prostate Cancer
Primary or Secondary Primary
Gene ID (Entrez) 3371.0
Test Specificity NB110-68136 specifically reacts with Domain B on FNIII repeats of Tenascin C.
Target Species Human, Mouse, Rat, Feline
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.