Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tenascin C Antibody (4C8MS), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB11068136PE
Description
Tenascin C Monoclonal specifically detects Tenascin C in Human, Mouse, Rat, Feline samples. It is validated for Flow Cytometry.Specifications
Tenascin C | |
Monoclonal | |
PE | |
P24821 | |
TNC | |
Recombinant human Tenascin C [Swiss-Prot# P24821]. | |
0.1 mL | |
Breast Cancer, Cancer, Extracellular Matrix, Neuroscience, Prostate Cancer | |
3371 | |
Human, Mouse, Rat | |
Purified |
Flow Cytometry | |
4C8MS | |
Flow Cytometry | |
150-225, Cytotactin, Glioma-associated-extracellular matrix antigen, GMEM, GP 150-225, Hexabrachion, hexabrachion (tenascin C, cytotactin), hexabrachion (tenascin), HXBcytotactin, JI, MGC167029, Myotendinous antigen, neuronectin, tenascin, tenascin C, Tenascin-C, tenascin-C isoform 14/AD1/16, TN-C, TNGP | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
NB110-68136 specifically reacts with Domain B on FNIII repeats of Tenascin C. | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction