Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tenascin C Antibody (SPM319), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP247920MFV610
Description
Tenascin C Monoclonal antibody specifically detects Tenascin C in Human, Rat (Negative) samples. It is validated for Flow CytometrySpecifications
Tenascin C | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Rat (Negative) | |
Purified |
Flow Cytometry | |
SPM319 | |
Flow Cytometry | |
150-225, Cytotactin, Glioma-associated-extracellular matrix antigen, GMEM, GP 150-225, Hexabrachion, hexabrachion (tenascin C, cytotactin), hexabrachion (tenascin), HXBcytotactin, JI, MGC167029, Myotendinous antigen, neuronectin, tenascin, tenascin C, Tenascin-C, tenascin-C isoform 14/AD1/16, TN-C, TNGP | |
Human breast carcinoma (Uniprot: P24821) | |
0.1 mL | |
Breast Cancer, Cancer, Cell Biology, Cellular Markers, Extracellular Matrix, Neuroscience, Prostate Cancer | |
3371 | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction