Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Tenascin C Monoclonal Antibody (4C8MS)
GREENER_CHOICE

Catalog No. PIMA516086
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA516086 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA516086 Supplier Invitrogen™ Supplier No. MA516086
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Tenascin C Monoclonal Antibody for Western Blot, IHC (P), IHC (F), ICC/IF, Flow, ELISA

Tenascin is a high molecular weight, multifunctional, extracellular matrix glycoprotein expressed in association with mesenchymal-epithelial interactions during development and in the neovasculature and stroma of undifferentiated tumors (Erickson and Bourdon 1989). It has been described under a variety of names: cytotactin, hexabrachion protein, J1, myotendinous antigen (MI) and glioma mesenchymal extracellular matrix (GMEM). The tenascin molecule is a disulfide-linked hexamer. The expression of tenascin is associated with development and growth, both normal and pathological, whereas the distribution in normal adult tissue is restricted.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tenascin C
Applications ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 4C8MS
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene TNC
Gene Accession No. P24821, Q80YX1
Gene Alias 150-225; AI528729; C130033P17Rik; cytotactin; deafness, autosomal dominant 56; DFNA56; Glioma-associated-extracellular matrix antigen; GMEM; GP; GP 150-225; hexabrachion; hexabrachion (tenascin); Hxb; JI; MGC144208; MGC144209; MGC167029; Myotendinous antigen; neuronectin; Ten; Tenascin; tenascin C; Tenascin-C; tenascin-C additional domain 1; tenascin-C isoform 14/AD1/16; TN; tnc; TN-C
Gene Symbols TNC
Host Species Mouse
Immunogen Recombinant human Tenascin C.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 101083629, 116640, 21923, 3371
Target Species Human, Mouse, Rat, Feline
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.