Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tenascin C Rabbit anti-Human,Mouse,Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA580140
Description
The synthetic peptide sequence is 27-52aa, NKYVQLNVGGSLYYTTVRALTRHDTM
In most tissues of adults, tenascin C is rarely present at significant levels. However, in developing tissues, or tissues experiencing a fast rate of growth, tenascin C is upregulated. This effect is particularly evident when this extracellular matrix glycoprotein is found in abundance in tissues responding to injury or infection. When muscular pH is increased, tenascin C production also increases. Prostate smooth muscle cells produce extra tenascin C when responding to LPS infection. In fibrosis and stenosis, tenascin C expression levels are markedly increased, and the protein seems to function as a mobilizer of immune cells responding to tissue damage and repair. In COPD, likewise, tenascin C expression is increased and is associated with cells attempting to repair damage within the lung. In short, tenascin C can be expected to be found in adult tissues only when associated with tissue damage and repair efforts.
Specifications
| Tenascin C | |
| Polyclonal | |
| PBS with 5MG BSA and 0.02% sodium azide, 0.02% thimerosal | |
| P24821, Q80YX1 | |
| Tnc | |
| A synthetic peptide corresponding to a sequence at the C-terminus of mouse Tenascin C. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| TNC | |
| Tenascin, TN, Hexabrachion, Tenascin-C, TN-C, Tnc | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 116640, 21923, 3371 | |
| -20°C | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction