Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tenascin C Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982765
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2765 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2765 Supplier Bioss Supplier No. BS1327R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Tenascin is a high molecular weight, multifunctional, extracellular matrix glycoprotein expressed in association with mesenchymal-epithelial interactions during development and in the neovasculature and stroma of undifferentiated tumors (Erickson and Bourdon 1989). It has been described under a variety of names: cytotactin, hexabrachion protein, J1, myotendinous antigen (MI) and glioma mesenchymal extracellular matrix (GMEM). The tenascin molecule is a disulfide-linked hexamer. The expression of tenascin is associated with development and growth, both normal and pathological, whereas the distribution in normal adult tissue is restricted.

Specifications

Antigen Tenascin C
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene TNC
Gene Accession No. P24821
Gene Alias 150-225; AI528729; C130033P17Rik; Cytotactin; deafness, autosomal dominant 56; DFNA56; Glioma-associated-extracellular matrix antigen; GMEM; GP; GP 150-225; Hexabrachion; hexabrachion (tenascin); Hxb; JI; MGC144208; MGC144209; MGC167029; Myotendinous antigen; Neuronectin; Ten; Tenascin; tenascin C; Tenascin-C; tenascin-C additional domain 1; tenascin-C isoform 14/AD1/16; TN; TNC; TN-C
Gene Symbols TNC
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Tenascin C.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3371
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.