Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tenascin R Monoclonal Antibody (1D12)
GREENER_CHOICE

Catalog No. PIMA520139
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA520139 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA520139 Supplier Thermo Scientific Supplier No. MA520139
Add to Cart
Add to Cart

Tenascin R Monoclonal Antibody (1D12)

Tenascin R Monoclonal Antibody for ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen Tenascin R
Applications ELISA
Classification Monoclonal
Clone 1D12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Tnr
Gene Accession No. Q92752
Gene Alias J1NRM; J1-tenascin; janusin; Neural recognition molecule J1-160/180; restrictin; tenascin R; tenascin R (restrictin, janusin); tenascin-R; Tenascin-R (Restrictin janusin J1-160/180); Tenascin-R (Restrictin, janusin, J1-160/180); Tnr; TN-R
Gene Symbols Tnr
Host Species Mouse
Immunogen TNR (NP_003276, 411 a.a. ~ 519 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7143
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.