Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Tenascin X Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144719
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144719 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144719 Supplier Invitrogen™ Supplier No. PA5144719
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PSSQLYEHTV EGGEKQVVFT HRINLPPSTG CGCPPGTEPP VLASEVQALR VRLEILEELV KGLKEQCTGG CCPASAQAGT GQTDVRTLCS LHGVFDLSRC TCSCEPGWGG PTCSDPTDA Antigen sequence identity: Mouse 92%; Rat 47%.

Tenascin-X (TNXB) is a extracellular matrix glycoprotein predominantly located in the outer reticular lamina of the basement membrane (BM). It interacts with many other ECM proteins and it accelerates collagen fibrillogenesisin vitro. TNXB plays a role in interactions between cell and ECM, antiadhesive effect, inhibiting cell migration, and in maintaining homeostasis of the extracellular matrix. Deficiency of TNXB has been associated with the connective tissue disorder Ehlers-Danlos syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tenascin X
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TNXB
Gene Accession No. P22105
Gene Alias EDS3; growth-inhibiting protein 45; hexabrachion-like protein; HXBL; tenascin XB; tenascin XB1; tenascin XB2; tenascin-X; TENX; Tn-mhc; Tnx; TN-X; TNXB; TNXB1; TNXB2; TNXB2tenascin XB1; TNXBS; TNXBSTNXTENX; VUR8; XB; XBS; XB-S; XBTN-X
Gene Symbols TNXB
Host Species Rabbit
Immunogen Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 55-173. Recombinant protein control fragment (Product #RP-109980).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7148
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.