Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Tenascin X Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104177
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104177 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104177 Supplier Invitrogen™ Supplier No. PA5104177
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total TNXB.

Tenascin-X (TNXB) is a extracellular matrix glycoprotein predominantly located in the outer reticular lamina of the basement membrane (BM). It interacts with many other ECM proteins and it accelerates collagen fibrillogenesisin vitro. TNXB plays a role in interactions between cell and ECM, antiadhesive effect, inhibiting cell migration, and in maintaining homeostasis of the extracellular matrix. Deficiency of TNXB has been associated with the connective tissue disorder Ehlers-Danlos syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tenascin X
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TNXB
Gene Accession No. P22105
Gene Alias EDS3; growth-inhibiting protein 45; hexabrachion-like protein; HXBL; tenascin XB; tenascin XB1; tenascin XB2; tenascin-X; TENX; Tn-mhc; Tnx; TN-X; TNXB; TNXB1; TNXB2; TNXB2tenascin XB1; TNXBS; TNXBSTNXTENX; VUR8; XB; XBS; XB-S; XBTN-X
Gene Symbols TNXB
Host Species Rabbit
Immunogen A synthesized peptide derived from human TNXB(Accession P22105), corresponding to amino acid residues L1745-S1795.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7148
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.