Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Tenax™ GR Thermal Desorption Tubes

Catalog No. 111005368 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Material:
Glass
Stainless Steel
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
11-100-5368 Glass
11-100-6695 Stainless Steel
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-100-5368 Supplier MilliporeSigma™ Supelco™ Supplier No. 29549U
Add to Cart
Add to Cart

Suitable for use with Air Monitoring

  • Fritted
  • Extends the range of Tenax™ TA
TRUSTED_SUSTAINABILITY

Specifications

Methods EPA TO-1, IP-1B
Diameter (Metric) Outer 6.35 mm
Includes Tenax™ GR Thermal Desorption Tube, TDS³ Storage Container
Overall Length (Metric) 88.9 mm
For Use With (Equipment) PerkinElmer, Markes, DANI, OI Analytical, and Shimadzu Instruments
Matrix Tenax GR Graphitized Porous Polymer
Product Line Tenax
For Use With (Application) Air Monitoring
Packaging Quantity 10/Pk.
Description Pre-conditioned
Diameter (English) Outer 1/4 in.
Overall Length (English) 3.5 in.
Material Glass
Type Thermal Desorption Tube
Show More Show Less

Tenax is a registered trademark of Buchem B.V.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.