Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Tenax™ GR Thermal Desorption Tubes

Catalog No. 111006695 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Material:
Glass
Stainless Steel
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
11-100-6695 Stainless Steel
11-100-5368 Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-100-6695 Supplier MilliporeSigma™ Supelco™ Supplier No. 20011U
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Suitable for use with Air Monitoring

  • Fritted
  • Extends the range of Tenax™ TA
TRUSTED_SUSTAINABILITY

Specifications

Methods EPA TO-1, IP-1B
Includes Tenax™ GR Thermal Desorption Tubes (10 ea.)
Diameter (Metric) Outer 6.35 mm
Overall Length (Metric) 88.9 mm
Material Stainless Steel
For Use With (Equipment) PerkinElmer, Markes, DANI, OI Analytical, and Shimadzu Instruments
For Use With (Application) Air Monitoring
Product Line Tenax
Matrix Tenax GR Graphitized Porous Polymer
Overall Length (English) 3.5 in.
Packaging Quantity 10/Pk.
Diameter (English) Outer 1/4 in.
Description Pre-conditioned
Type Thermal Desorption Tube
Show More Show Less

Tenax is a registered trademark of Buchem B.V.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.