Learn More
Sigma Aldrich Tenax PC-ABS 3D Printing Filament
Catalog No. 503126066 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
50-312-6066 1 Ea.
1 options
Catalog No. 50-312-6066 Supplier Sigma Aldrich Supplier No. TRD3D00601EA
Only null left
Add to Cart
Add to Cart
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
ParametersExtruder (nozzle) temperature: 250 +/- 10°CBed temperature: 100 +/- 10°CPrint speed: 50-60 mm/sFan: 0-20 %Minimum nozzle diameter: 0.4 mmPlate materials: blue tape, kapton tape, etc.
Specifications
Quantity 1 Ea.