Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Tenax™ TA Thermal Desorption Tubes

Catalog No. 111005358 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Description:
Pre-conditioned
Pre-conditioned/Sealed with Brass Endcaps
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
11-100-5358 Pre-conditioned
11-100-5349 Pre-conditioned/Sealed with Brass Endcaps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-100-5358 Supplier MilliporeSigma™ Supelco™ Supplier No. 29530U
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Suitable for use with Air Monitoring

  • Fritted
  • Particle Size: 35/60 Mesh
TRUSTED_SUSTAINABILITY

Specifications

Includes Tenax™ TA Thermal Desorption Tubes (10)
Diameter (Metric) Outer 6.35 mm
Overall Length (Metric) 88.9 mm
Packaging Quantity 10/Pk.
Methods EPA TO-1, IP-1 B
For Use With (Application) Air Monitoring
Material Glass
Product Line Tenax
Matrix Tenax™ TA Porous Polymer (35/60)
Overall Length (English) 3.5 in.
Diameter (English) Outer 1/4 in.
Description Pre-conditioned
For Use With (Equipment) PerkinElmer, Markes, DANI, OI Analytical, and Shimadzu Instruments
Type Thermal Desorption Tube
Show More Show Less

Tenax is a registered trademark of Buchem B.V.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.