Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Tenax™ TA Thermal Desorption Tubes

Catalog No. p-7202933 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Description:
Pre-conditioned
Pre-conditioned/Sealed with Brass Endcaps
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
11-100-5365 Pre-conditioned
11-100-5350 Pre-conditioned/Sealed with Brass Endcaps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-100-5365 Supplier MilliporeSigma™ Supelco™ Supplier No. 29539U
Only null left
Add to Cart
Add to Cart

Suitable for use with Air Monitoring

  • Fritted
  • Particle Size: 60/80 Mesh
TRUSTED_SUSTAINABILITY

Specifications

Product Line Tenax
Material Glass
For Use With (Application) Air Monitoring
Methods EPA TO-1, IP-1 B
Packaging Quantity 10/Pk.
Diameter (English) Outer 1/4 in.
Overall Length (English) 3 1/2 in.
Description Pre-conditioned
Matrix Tenax™ TA Porous Polymer (60/80)
For Use With (Equipment) PerkinElmer, Markes, DANI, OI Analytical, and Shimadzu Instruments
Type Thermal Desorption Tube
Show More Show Less

Tenax is a registered trademark of Buchem B.V.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.