Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TENC1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA557006
Description
Immunogen sequence: VPSQMPWLVA SPEPPQSSPT PAFPLAASYD TNGLSQPPLP EKRHLPGPGQ QPGPWGPEQA SSPARGISHH VTFAPLLSDN VPQTPEPPTQ ESQSN Highest antigen sequence identity to the following orthologs: Mouse - 69%, Rat - 68%.
Regulates cell motility and proliferation. May have phosphatase activity. Reduces AKT1 phosphorylation. Lowers AKT1 kinase activity and interferes with AKT1 signaling.
Specifications
| TENC1 | |
| Polyclonal | |
| Unconjugated | |
| TNS2 | |
| C1 domain-containing phosphatase and tensin homolog; C1 domain-containing phosphatase and tensin-like protein; C1TEN; C1-ten; KIAA1075; nep; nph; Tenc1; tensin 2; tensin like C1 domain containing phosphatase (tensin 2); tensin like C1 domain-containing phosphatase; tensin-2; Tensin-like C1 domain-containing phosphatase; Tns2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 23371 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q63HR2 | |
| TNS2 | |
| Recombinant protein corresponding to Human TENC1. Recombinant protein control fragment (Product #RP-96517). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction