Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TENC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103174
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103174 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103174 Supplier Invitrogen™ Supplier No. PA5103174
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total Tensin-2.

Regulates cell motility and proliferation. May have phosphatase activity. Reduces AKT1 phosphorylation. Lowers AKT1 kinase activity and interferes with AKT1 signaling.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TENC1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TNS2
Gene Accession No. Q63HR2, Q8CGB6
Gene Alias C1 domain-containing phosphatase and tensin homolog; C1 domain-containing phosphatase and tensin-like protein; C1TEN; C1-ten; KIAA1075; nep; nph; Tenc1; tensin 2; tensin like C1 domain containing phosphatase (tensin 2); tensin like C1 domain-containing phosphatase; tensin-2; Tensin-like C1 domain-containing phosphatase; Tns2
Gene Symbols TNS2
Host Species Rabbit
Immunogen A synthesized peptide derived from human TNS2(Accession Q63HR2), corresponding to amino acid residues P968-Q1018.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 209039, 23371
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.