Learn More
Invitrogen™ TENM3 Polyclonal Antibody

Description
The antibody detects endogenous levels of total ODZ3 / Teneurin-3 protein.
Specifications
Specifications
| Antigen | TENM3 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1.7 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | TENM3 |
| Gene Accession No. | Q9P273, Q9WTS6 |
| Gene Alias | 2610100B16Rik; FLJ10474; FLJ10886; KIAA1455; MCOPCB9; mKIAA1455; odd Oz/ten-m homolog 3; odz, odd Oz/ten-m homolog 3; Odz1; Odz3; ODZ3-like protein; protein Odd Oz/ten-m homolog 3; Ten-3; Tenascin-M3; teneurin transmembrane protein 3; Teneurin-3; TENM3; Ten-m3; testicular tissue protein Li 195; TNM3 |
| Gene Symbols | TENM3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.