Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Tenofovir
GREENER_CHOICE

Catalog No. 366610R
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
36-661-0R 10 mg
36-665-0R 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 36-661-0R Supplier Tocris Bioscience™ Supplier No. 3666/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Reverse transcriptase inhibitor

Tenofovir is a selectively inhibits HIV reverse transcriptase (RNA-dependent DNA polymerase). Prevents cytotoxicity in SIV-infected C-8166 cells in vitro (IC50 = 1.5 μM). Antiviral agent.

Specifications

CAS 147127-20-6
Quantity 10 mg
Molecular Formula (R)-[[2-(6-Amino-9 H-purin-9-yl)-1-methylethoxy]methyl]phosphonic acid
Product Type Tenofovir
Purity 99%
Molecular Weight (g/mol) M.W. (Observed): 287.21
Inhibitors Tenofovir

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.