Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tenofovir disoproxil fumarate, 98%, Thermo Scientific Chemicals

Catalog No. ac461250010
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 202138-50-9
Molecular Formula C19H30N5O10P·C4H4O4
Molecular Weight (g/mol) 635.51
InChI Key VCMJCVGFSROFHV-WZGZYPNHSA-N
Synonym tenofovir disoproxil fumarate, viread, tenofovir df, unii-ott9j7900i, tenofovir disoproxil fumarate usan, 9-r-2-bis isopropoxycarbonyl oxy methoxy phosphinyl methoxy propyl adenine fumarate, 9-r-2-bis isopropoxycarbonyl oxy methoxy phosphinyl methoxy propyl adenine, fumarate, r-5-2-6-amino-9h-purin-9-yl-1-methylethoxy methyl-2,4,6,8-tetraoxa-5-phosphanonanedioic acid, bis 1-methylethyl ester, 5-oxide, e-2-butenedioate 1:1, 9-r-2-bis isopropoxycarbonyl oxy methoxy phosphinyl methoxy propyl adenine fumarate 1:1
PubChem CID 6398764
ChEBI CHEBI:63718
IUPAC Name [[(2R)-1-(6-aminopurin-9-yl)propan-2-yl]oxymethyl-(propan-2-yloxycarbonyloxymethoxy)phosphoryl]oxymethyl propan-2-yl carbonate;(E)-but-2-enedioic acid
SMILES CC(C)OC(=O)OCOP(=O)(COC(C)CN1C=NC2=C1N=CN=C2N)OCOC(=O)OC(C)C.C(=CC(=O)O)C(=O)O

Specifications

Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Solubility Information Solubility in water: >10 mg/mL.
Formula Weight 635.51
Percent Purity 98%
Chemical Name or Material Tenofovir disoproxil fumarate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.