Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tensin 1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 5 publications
$416.50 - $670.00
Specifications
Antigen | Tensin 1 |
---|---|
Dilution | Western Blot Reported in scientific literature (PMID: 30813060)., Immunohistochemistry 1:20 - 1:50, Immunocytochemistry/Immunofluorescence Reactivity reported in (PMID: 25766369). , Immunohistochemistry-Paraffin 1:20 - 1:50 |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Tensin 1 Polyclonal specifically detects Tensin 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Tensin 1 | |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human, Mouse, Rat | |
DKFZp586K0617, matrix-remodelling associated 6, Matrix-remodelling-associated protein 6, MGC88584, MST091, MST122, MST127, MSTP127, MXRA6, tensin, tensin 1, tensin-1, TNSMSTP122 | |
TNS1 | |
IgG | |
Affinity Purified | |
Specificity of human Tensin 1 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Western Blot Reported in scientific literature (PMID: 30813060)., Immunohistochemistry 1:20 - 1:50, Immunocytochemistry/Immunofluorescence Reactivity reported in (PMID: 25766369). , Immunohistochemistry-Paraffin 1:20 - 1:50 | |
Polyclonal | |
Rabbit | |
Cell Cycle and Replication, Signal Transduction | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
7145 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:EEPLNLEGLVAHRVAGVQAREKQPAEPPAPLRRRAASDGQYENQSPEATSPRSPGVRSPVQCVSPELALTIALNPGGRPKEPHLHSYK | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title