Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tensin 1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $646.00
Specifications
Antigen | Tensin 1 |
---|---|
Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), KnockDown |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Tensin 1 Polyclonal specifically detects Tensin 1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Knockdown Validated.Specifications
Tensin 1 | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), KnockDown | |
Unconjugated | |
RUO | |
Human | |
DKFZp586K0617, matrix-remodelling associated 6, Matrix-remodelling-associated protein 6, MGC88584, MST091, MST122, MST127, MSTP127, MXRA6, tensin, tensin 1, tensin-1, TNSMSTP122 | |
TNS1 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Western Blot 0.4 ug/ml, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
Polyclonal | |
Rabbit | |
Cell Cycle and Replication, Signal Transduction | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
7145 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:LSPLTNGLDKSYPMEPMVNGGGYPYESASRAGPAHAGHTAPMRPSYSAQEGLAGYQREGPHPAWPQPVTTSHYAHDPSGMFRSQS | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title