Learn More
Invitrogen™ Tensin 1 Polyclonal Antibody

Description
Predicted to react with Mouse samples.
Specifications
Specifications
| Antigen | Tensin 1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | TNS1 |
| Gene Accession No. | Q9HBL0 |
| Gene Alias | 1110018I21Rik; 1200014E20Rik; AI648117; DKFZp586K0617; E030018G17Rik; E030037J05Rik; matrix-remodelling associated 6; Matrix-remodelling-associated protein 6; MGC88584; MST091; MST122; MST127; MSTP091; MSTP122; MSTP127; MXRA6; PPP1R155; protein phosphatase 1, regulatory subunit 155; tensin 1; tensin-1; TNS; TNS1 |
| Gene Symbols | TNS1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.