Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Tensin 1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116023
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116023 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116023 Supplier Invitrogen™ Supplier No. PA5116023
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total Tensin-1.

Tensin is involved in the maintenance of cellular structure by anchoring Actin filaments at the focal adhesion via F-Actin binding and capping activities. However, tensin also contains a Src homology 2 (SH2) domain and has the ability to be phosphorylated. Tensin is phosphorylated on tyrosine, serine, and threonine residues, suggesting that it might participate in signal transduction cascades. These diverse characteristics in a single molecule indicate that tensin may be an important link between the cytoskeleton and signal transduction pathways.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tensin 1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene TNS1
Gene Accession No. Q9HBL0
Gene Alias 1110018I21Rik; 1200014E20Rik; AI648117; DKFZp586K0617; E030018G17Rik; E030037J05Rik; matrix-remodelling associated 6; Matrix-remodelling-associated protein 6; MGC88584; MST091; MST122; MST127; MSTP091; MSTP122; MSTP127; MXRA6; PPP1R155; protein phosphatase 1, regulatory subunit 155; tensin 1; tensin-1; TNS; TNS1
Gene Symbols TNS1
Host Species Rabbit
Immunogen A synthesized peptide derived from human TNS1(Accession Q9HBL0), corresponding to amino acid residues N1651-T1701.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21961, 7145
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.