Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Tensin 3 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA563238

Catalog No. PIPA563238


Only null left
Add to Cart

Description

Description

Immunogen sequence: VGSGPHPPDT QQPSPSKAFK PRFPGDQVVN GAGPELSTGP SPGSPTLDID QSIEQLNRLI LELDPTFEPI PTHMNALGSQ ANGSV Highest antigen sequence identity to the following orthologs: Mouse - 69%, Rat - 73%.

The Tensin (Tns) family of proteins is involved in the maintenance of cellular structure by anchoring actin filaments at the focal adhesion via F-actin binding and capping activities. Tns proteins also contain a Src homology 2 (SH2) domain and have the ability to be phosphorylated, suggesting a role in signal transduction cascades. These diverse characteristics indicate that Tns proteins may be important links between the cytoskeleton and signal transduction pathways. Tns3, also known as TEM6 or TENS1, localizes to the focal adhesions of the plasma membrane. It is predominantly expressed in thyroid and placenta but can also be found in heart, liver, brain, prostate, pancreas, kidney, lung, skeletal muscle and white blood cells. Tns3 is essential for proper growth and development, as suggested by growth retardation and death in a number of Tns3-/- mice.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Tensin 3
Polyclonal
Unconjugated
Tns3
BC023928; F830010I22Rik; RGD1564174; TEM6; TENS1; TENS3; tensin 3; tensin3; tensin-3; tensin-3-like; tensin-like SH2 domain containing 1; tensin-like SH2 domain-containing 1; tensin-like SH2 domain-containing protein 1; thyroid specific PTB domain protein; TNS3; TPP; TSN3; tumor endothelial marker 6
Rabbit
Antigen affinity chromatography
RUO
64759
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Immunocytochemistry
0.3 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q68CZ2
Tns3
Recombinant protein corresponding to Human Tensin 3. Recombinant protein control fragment (Product #RP-103240).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.