Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Tensin 3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570772
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570772 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570772 Supplier Invitrogen™ Supplier No. PA570772
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 86%; Dog: 100%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.

The Tensin (Tns) family of proteins is involved in the maintenance of cellular structure by anchoring actin filaments at the focal adhesion via F-actin binding and capping activities. Tns proteins also contain a Src homology 2 (SH2) domain and have the ability to be phosphorylated, suggesting a role in signal transduction cascades. These diverse characteristics indicate that Tns proteins may be important links between the cytoskeleton and signal transduction pathways. Tns3, also known as TEM6 or TENS1, localizes to the focal adhesions of the plasma membrane. It is predominantly expressed in thyroid and placenta but can also be found in heart, liver, brain, prostate, pancreas, kidney, lung, skeletal muscle and white blood cells. Tns3 is essential for proper growth and development, as suggested by growth retardation and death in a number of Tns3-/- mice.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tensin 3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Tns3
Gene Accession No. Q5SSZ5
Gene Alias BC023928; F830010I22Rik; RGD1564174; TEM6; TENS1; TENS3; tensin 3; tensin3; tensin-3; tensin-3-like; tensin-like SH2 domain containing 1; tensin-like SH2 domain-containing 1; tensin-like SH2 domain-containing protein 1; thyroid specific PTB domain protein; TNS3; TPP; TSN3; tumor endothelial marker 6
Gene Symbols Tns3
Host Species Rabbit
Immunogen synthetic peptide directed towards the C-terminal region of mouse Tensin 3.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 319939
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.