Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Tensin 4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5116288
Description
Tensin 4 Polyclonal Antibody for Western Blot
TNS4 links the focal adhesion and cell adhesion signaling to the actin cytoskeleton. It may be involved in cell survival and migration. The protein may be involved in cell migration, cartilage development and in linking signal transduction pathways to the cytoskeleton and promote apoptosis, via its cleavage by caspase-3.
Specifications
Tensin 4 | |
Polyclonal | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q8IZW8 | |
TNS4 | |
KLH conjugated synthetic peptide between 132-159 amino acids from the N-terminal region of human CTEN. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
TNS4 | |
9930017A07Rik; AA589547; AU016405; C terminal tensin like; CTEN; c-ten; C-terminal tensin-like protein; PP14434; RGD1310402; tensin 4; tensin-4; TNS4 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
84951 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction