Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Tensin 4 Recombinant Rabbit Monoclonal Antibody (PD00-95)
GREENER_CHOICE

Catalog No. FEMA550733
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550733 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550733 Supplier Invitrogen™ Supplier No. MA550733
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive control: Human liver carcinoma tissue, human placenta tissue. Predicted band size: 77 kDa Subcellular Location: Cell junction, focal adhesion, Cytoplasm, cytoskeleton.

TNS4 links the focal adhesion and cell adhesion signaling to the actin cytoskeleton. It may be involved in cell survival and migration. The protein may be involved in cell migration, cartilage development and in linking signal transduction pathways to the cytoskeleton and promote apoptosis, via its cleavage by caspase-3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tensin 4
Applications Immunohistochemistry (Paraffin)
Classification Recombinant Monoclonal
Clone PD00-95
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene TNS4
Gene Accession No. Q8IZW8
Gene Alias 9930017A07Rik; AA589547; AU016405; C terminal tensin like; CTEN; c-ten; C-terminal tensin-like protein; PP14434; RGD1310402; tensin 4; tensin-4; TNS4
Gene Symbols TNS4
Host Species Rabbit
Immunogen Synthetic peptide within Human TNS4/CTEN aa 700 to the C-terminus (C terminal).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84951
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.