Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tentonin 3/TMEM150c, Rabbit anti-Mouse, Rat, Polyclonal, Millipore Sigma™

Catalog No. ABN226625UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABN226625UL 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABN226625UL Supplier MilliporeSigma Supplier No. ABN226625UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Tentonin 3/TMEM150c, Cat. No. ABN2266, is a highly specific rabbit polyclonal antibody that targets Transmembrane protein 150C and has been tested in , Immunocytochemistry, Immunofluorescence, Immunohistochemistry, and Western Blotting.

Transmembrane protein 150C (UniProt: Q8C8S3; also known as Tentonin, TTN3) is encoded by the Tmem150c gene (Gene ID: 231503) in murine species. TTN3 is a multi-pass membrane protein of the DRAM/TMEM150 family. Two isoforms of TTN3 have been described that are produced by alternative splicing. It belongs to a new class of mechanotransduction channel that is expressed in muscle spindle afferents and contribute to proprioception (touch sensation). It is activated by mechanical stimuli with distinctly slow inactivation kinetics. Mice with deficiency in TTN3 display loss of coordinated movements and abnormal gait. It is reported to mediate slow adapting currents in DRG neurons.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tentonin 3/TMEM150c
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry Analysis: A representative lot detected Tentonin 3/TMEM150c in muscle spindle from mouse (Hong, G.S., et. al. (2016). Neuron. 91(1):107-18). Immunocytochemistry Analysis: A representative lot detected Tentonin 3/TMEM150c in HEK293T cells transfected with TTN3. (Hong, G.S., et. al. (2016). Neuron. 91(1):107-18).Western Blotting Analysis: A representative lot detected Tentonin 3/TMEM150c in F11 rat DRG cells (untransfected and TTN3 transfected cells) (Hong, G.S., et. al. (2016). Neuron. 91(1):107-18).
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Transmembrane protein 150C;TTN3;Tentonin 3
Gene Symbols Tmem150c
Host Species Rabbit
Immunogen A linear peptide corresponding to 38 amino acids from the C-terminal region of murine Transmembrane protein 150C.
Molecular Weight of Antigen ∽25 kDa observed; 27.76 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Test Specificity This rabbit polyclonal antibody detects Transmembrane protein 150C in mouse and rat cells. it targets an epitope within 38 amino acids from the C-terminal region.
Target Species Mouse, Rat
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.