Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Teplizumab Antibody (2505B), Allophycocyanin/Cy7, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals FAB10245APCCY7
Description
Teplizumab Monoclonal antibody specifically detects Teplizumab in Broad Species Reactivity samples. It is validated for Flow CytometrySpecifications
Teplizumab | |
Monoclonal | |
APC-Cyanine7 | |
PBS | |
Full-length Teplizumab | |
0.1 mL | |
Primary | |
Store at 4C in the dark. Do not freeze. | |
IgG |
Flow Cytometry | |
2505B | |
Flow Cytometry | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
RUO | |
Broad Species Reactivity | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction