Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Teprotumumab Recombinant Human Monoclonal Antibody
GREENER_CHOICE

Catalog No. PIMA541915
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA541915 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA541915 Supplier Invitrogen™ Supplier No. MA541915
Add to Cart
Add to Cart
This item is not returnable. View return policy

Human Recombinant Monoclonal Antibody

Endotoxin level < 0.01EU/μg by LAL method Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.

Teprotumumab is a fully human IgG1 monoclonal antibody directed against the human insulin-like growth factor-1 receptor. 8 Following a clinical trial in which its efficacy in the treatment of thyroid eye disease (TED) was assessed, it received 'breakthrough therapy' designation from the FDA in 20163 and was approved by the FDA in January 2020 for the treatment of TED. 7 Thyroid eye disease is a potentially debilitating complication of Graves' Disease involving inflammation and tissue remodeling behind the eye, and previous treatment options typically involved multiple invasive surgeries - teprotumumab is the first pharmaceutical ever approved for the treatment of TED and therefore represents a significant step forward in the treatment this disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Teprotumumab
Applications ELISA, SDS-Page
Classification Recombinant Monoclonal
Concentration 1.74 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Alias RO4858696-000
Host Species Human
Immunogen Human IGF1R/CD221.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Target Molecule Chemical
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.