Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TER-119 Antibody (TER-119), Allophycocyanin/Cy7, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB1125APCCY7
Description
TER-119 Monoclonal antibody specifically detects TER-119 in Mouse samples. It is validated for Flow CytometrySpecifications
TER-119 | |
Monoclonal | |
APC-Cyanine7 | |
PBS | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
104231 | |
Store at 4C in the dark. Do not freeze. | |
IgG2b |
Flow Cytometry | |
TER-119 | |
Flow Cytometry | |
Ly76, Ly-76, TER119 | |
C57BL/6 mouse day-14 fetal liver cells | |
0.5 mL | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction