Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TER-119 Antibody (TER-119), HRP, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB1125H
This item is not returnable.
View return policy
Description
TER-119 Monoclonal antibody specifically detects TER-119 in Mouse samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
TER-119 | |
Monoclonal | |
HRP | |
Ly76, Ly-76, TER119 | |
C57BL/6 mouse day-14 fetal liver cells | |
0.5 mL | |
Primary | |
Mouse | |
Purified |
Western Blot, Immunohistochemistry | |
TER-119 | |
PBS | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
104231 | |
Store at 4°C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction