Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TER-119/Erythroid Cells Rat anti-Mouse, Brilliant Violet 650, Clone: TER-119, BD OptiBuild™

Catalog No. BDB747739 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB747739 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB747739 Supplier BD Biosciences Supplier No. 747739
Add to Cart
Add to Cart

Rat Monoclonal Antibody

TER-119/Erythroid Cells Monoclonal antibody specifically detects TER-119/Erythroid Cells in Mouse samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TER-119/Erythroid Cells
Applications Flow Cytometry
Classification Monoclonal
Clone TER-119
Concentration 0.2 mg/mL
Conjugate Brilliant Violet 650
Host Species Rat
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity TER-119/Erythroid Cells
Target Species Mouse
Content And Storage Refrigerate (2 to 8°C)
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.