Learn More
TER-119/Erythroid Cells Rat anti-Mouse, BUV496, Clone: TER-119, BD Biosciences™
Shop All BD Biosciences ProductsDescription
Specifications
Specifications
| Antigen | TER-119/Erythroid Cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | TER-119 |
| Conjugate | Brilliant Ultraviolet 496 |
| Formulation | Aqueous buffered solution containing ≤0.09% sodium azide. |
| Gene Alias | Lymphocyte antigen 76; Ly76; Ly-76; TER-119; Ter119 |
| Host Species | Rat |
| Immunogen | Mouse Fetal Liver |
| Quantity | 50 μg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.