Learn More
TER-119/Erythroid cells Rat anti-Mouse, FITC, Clone: TER-119, BD
Rat Monoclonal Antibody
$74.00 - $154.70
Specifications
Antigen | TER-119/Erythroid cells |
---|---|
Clone | TER-119 |
Concentration | 0.5mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
The TER-119 antibody specifically binds to a 52 kDa molecule associated with glycophorin A on cells of the erythroid lineage in embryonic yolk sac, fetal liver, newborn liver, adult bone marrow, adult peripheral blood, and adult lymphoid organs. The TER-119 antigen is expressed on erythroid cells from pro-erythroblast through mature erythrocyte stages, but not on cells with BFU-E or CFU-E activities. The TER-119 epitope is not detected on hematopoietic stem cells, lymphoid cells, myeloid cells, or erythroleukemia lines. The TER-119 mAb is a component of the “lineage cocktail” used in studies of hematopoietic progenitors to detect, or deplete cells committed to the hematopoietic lineages.
Specifications
TER-119/Erythroid cells | |
0.5mg/mL | |
Monoclonal | |
Rat | |
Mouse | |
Mouse Fetal Liver | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
TER-119 | |
Flow Cytometry | |
FITC | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
IgG2b κ | |
Affinity Purified | |
Lymphocyte antigen 76; Ly76; Ly-76; TER-119; Ter119 |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.