Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TER-119/Erythroid Cells Rat anti-Mouse, PE, Clone: TER-119, BD

Catalog No. BDB553673 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.2 mg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB553673 0.2 mg
BDB561071 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB553673 Supplier BD Biosciences Supplier No. 553673
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The TER-119 antibody reacts with a 52kDa molecule associated with glycophorin A on cells of the erythroid lineage in embryonic yolk sac, fetal liver, newborn liver, adult bone marrow, adult peripheral blood, and adult lymphoid organs. The TER-119 antigen is expressed on erythroid cells from pro-erythroblast through mature erythrocyte stages, but not on cells with BFU-E or CFU-E activities. The TER-119 epitope is not detected on hematopoietic stem cells, lymphoid cells, myeloid cells, or erythroleukemia lines. The TER-119 mAb is a component of the “lineage cocktail” used in studies of hematopoietic progenitors to detect, or deplete cells committed to the hematopoietic lineages. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Host Species: Rat
Clone: TER-119
Isotype: IgG2b, κ
Species Reactivity: Mouse
Immunogen: Mouse Fetal Liver

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen TER-119/Erythroid Cells
Applications Flow Cytometry
Classification Monoclonal
Clone TER-119
Concentration 0.2mg/mL
Conjugate PE
Description Lymphocyte antigen 76; Ly76; Ly-76; TER-119; Ter119
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Mouse Fetal Liver
Purification Method Affinity Purified
Quantity 0.2 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2b κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.