Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TER-119 Monoclonal Antibody (TER-119), Biotin

Rat Monoclonal Antibody
$326.00 - $630.00
Specifications
| Antigen | TER-119 |
|---|---|
| Clone | TER-119 |
| Concentration | 0.1 mg/mL |
| Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| Applications | Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA517819
![]() |
Invitrogen™
MA517819 |
100 μg |
Each for $326.00
|
|
|||||
|
PIMA517820
|
Invitrogen™
MA517820 |
300 μg |
Each for $630.00
|
|
|||||
Description
TER-119 Monoclonal Antibody for IHC (F), Flow, IP
TER-119 is a lineage marker for erythroid cells from early proerythroblast to mature erythrocyte stages in adult blood, spleen, and bone marrow. It is also present in yolk sac, and fetal and newborn liver. The TER-119 antigen is not expressed by cells of earlier erythroid development at BFU-e (blast-forming unit erythroid) stage or CFU-e (colony-forming unit erythroid) stage.Specifications
| TER-119 | |
| 0.1 mg/mL | |
| Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation | |
| Biotin | |
| Rat | |
| RUO | |
| PBS with 4-5mg/mL BSA and 0.02% sodium azide | |
| 0 | |
| 104231 | |
| Day 14 BALB/c fetal liver cells. | |
| Antibody |
| TER-119 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Monoclonal | |
| Liquid | |
| IgG2b | |
| Mouse | |
| Ly76 | |
| Ly76; Ly-76; TER119 | |
| Ly76 | |
| Primary | |
| Protein G |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title